Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-chloro-6-methyl-4-{[(1R)-1-(1-methylpiperidin-4-yl)ethyl]sulfamoyl}phenyl)-2-methylbenzamide | 2756350-98-6 | MFCD00004280 | 98.3% | 464.02 Da | C23H30ClN3O3S | 5 MG
SDP

Catalog No. 5000222706
Encompass_Preferred

CCR8 antagonist 2 is a small-molecule CCR8 receptor antagonist used as a research tool to probe CCR8-mediated signaling in immunology and inflammation studies. It is routinely applied in biochemical and cellular assays to study regulatory T cell and Th2 responses.

  • High purity confirmed by HPLC (98.3%).
  • Potent CCR8 antagonist activity for receptor-targeted studies.
  • Suitable for biochemical and cellular CCR8 assays.
  • Soluble in DMSO for easy preparation of stock solutions.
  • Molecular weight 464.02 Da; molecular formula C23H30ClN3O3S.
  • Stable when stored at -20 °C for long-term storage.

Catalog No. 50-002-22706 Supplier Medchemexpress LLC Supplier No. HY1442005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCR8 antagonist 2 is a small-molecule CCR8 receptor antagonist used as a research tool to probe CCR8-mediated signaling in immunology and inflammation studies. It is routinely applied in biochemical and cellular assays to study regulatory T cell and Th2 responses.

  • High purity confirmed by HPLC (98.3%).
  • Potent CCR8 antagonist activity for receptor-targeted studies.
  • Suitable for biochemical and cellular CCR8 assays.
  • Soluble in DMSO for easy preparation of stock solutions.
  • Molecular weight 464.02 Da; molecular formula C23H30ClN3O3S.
  • Stable when stored at -20 °C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.