Learn More
Medchemexpress LLC N-(2-chlorophenyl)-6-(piperidin-4-yl)imidazo[1,2-a]pyridine-3-carboxamide oxalate | 1198408-78-4 | MFCD28023573 | 99.0% | 444.87 g/mol | C21H21ClN4O5 | 1 MG
SDP
Catalog No. 5000240341
Encompass_Preferred
LDN-211904 oxalate is the oxalate salt of a research small molecule described as a potent, reversible EphB3 inhibitor for biochemical and cellular studies. It is provided in milligram quantities for laboratory research, with a reported molecular weight of 444.87 g/mol and CAS 1198408-78-4. Not for clinical or human use.
  • Potent, reversible EphB3 inhibitor (IC50 ≈ 79 nM).
  • Reported molecular weight 444.87 g/mol and CAS 1198408-78-4.
  • High purity suitable for research (>99.0%).
  • Demonstrated metabolic stability in mouse liver microsomes.
  • Supplied in research-scale quantities (for example, 1 mg).
Catalog No. 50-002-40341 Supplier Medchemexpress LLC Supplier No. HY1074601MG
LDN-211904 oxalate is the oxalate salt of a research small molecule described as a potent, reversible EphB3 inhibitor for biochemical and cellular studies. It is provided in milligram quantities for laboratory research, with a reported molecular weight of 444.87 g/mol and CAS 1198408-78-4. Not for clinical or human use.
  • Potent, reversible EphB3 inhibitor (IC50 ≈ 79 nM).
  • Reported molecular weight 444.87 g/mol and CAS 1198408-78-4.
  • High purity suitable for research (>99.0%).
  • Demonstrated metabolic stability in mouse liver microsomes.
  • Supplied in research-scale quantities (for example, 1 mg).