Learn More
Medchemexpress LLC N-(2-cyclopropyl-7-fluoro-4-oxoquinazolin-3(4H)-yl)-2-(4-fluorophenyl)ethanamide | 2694728-63-5 | 99.9% | 355.34 | C19H15F2N3O2 | 100MG
Supplier: Medchemexpress LLC HY125469100MG
ICA-105665 (PF-04895162) is a small-molecule opener of neuronal Kv7 potassium channels supplied as a solid for laboratory research use. It crosses the blood-brain barrier, has demonstrated broad-spectrum antiseizure activity in animal models, and displays BSEP inhibition at high concentrations. Store and handle according to solubility and storage recommendations for research compounds.