Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-fluoro-4-((2-(6-methoxy-1,6-dihydropyridin-3-yl)pyrimidin-4-yl)oxy)phenyl)-1,5-dimethyl-3-oxo-2 | 888719-03-7 | 99.5% | 458.42 g·mol⁻¹ | C25H16F2N4O3 | 10 MG
SDP

Catalog No. 5000295336
Encompass_Preferred

MET kinase-IN-4 is a potent, orally active inhibitor of the MET receptor tyrosine kinase intended for research use. It exhibits low-nanomolar MET inhibition and activity against FLT3 and VEGFR-2, supplied as a solid for in vitro and in vivo studies.

  • Potent MET inhibition (IC50 1.9 nM).
  • Additional activity against FLT3 (IC50 4 nM) and VEGFR-2 (IC50 27 nM).
  • High purity: 99.5%.
  • Solid, off-white to light yellow appearance.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended.
  • Storage: 4°C sealed; in solution -80°C for 6 months, -20°C for 1 month.
  • Molecular formula C25H16F2N4O3; molecular weight 458.42 g·mol⁻¹.

Catalog No. 50-002-95336 Supplier Medchemexpress LLC Supplier No. HY1830910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MET kinase-IN-4 is a potent, orally active inhibitor of the MET receptor tyrosine kinase intended for research use. It exhibits low-nanomolar MET inhibition and activity against FLT3 and VEGFR-2, supplied as a solid for in vitro and in vivo studies.

  • Potent MET inhibition (IC50 1.9 nM).
  • Additional activity against FLT3 (IC50 4 nM) and VEGFR-2 (IC50 27 nM).
  • High purity: 99.5%.
  • Solid, off-white to light yellow appearance.
  • Soluble in DMSO at 100 mg/mL; ultrasonic recommended.
  • Storage: 4°C sealed; in solution -80°C for 6 months, -20°C for 1 month.
  • Molecular formula C25H16F2N4O3; molecular weight 458.42 g·mol⁻¹.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.