Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Fluorophenyl)-3-oxobutyramide 98.0+%, TCI America™
SDP

Catalog No. F09631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09631G 1 g
F0963200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09631G Supplier TCI America Supplier No. F09631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5279-85-6
Molecular Formula C10H10FNO2
Molecular Weight (g/mol) 195.19
MDL Number MFCD00661533
InChI Key SNNJOLBZQNBODQ-UHFFFAOYSA-N
Synonym 2′C-Fluoroacetoacetanilide
PubChem CID 763281
IUPAC Name N-(2-fluorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1F

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 1 g
Formula Weight 195.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Fluorophenyl)-3-oxobutyramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.