Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[2-hydroxy-3-(1-piperidinyl)propoxy]-3-pyridinecarboximidamide dihydrochloride | 66611-37-8 | 98.0% | 351.27 g/mol | C14H24Cl2N4O2 | 1 ML
SDP

Catalog No. 5000211727
Encompass_Preferred

BGP-15 is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor for research use, characterized in the literature for cytoprotective and insulin-sensitizing effects. It is available as a ready-to-use solution or as solid material and is commonly used to probe PARP-related pathways in biochemical and cellular studies.

  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • Demonstrates PARP inhibition with IC50 = 120 μM and Ki = 57 μM.
  • High purity, 98.0% by HPLC.
  • Molecular weight approximately 351.27 g/mol.
  • CAS number 66611-37-8 for unambiguous identification.
  • Suitable for biochemical and cell-based research applications.

Catalog No. 50-002-11727 Supplier Medchemexpress LLC Supplier No. HY10082810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BGP-15 is a small-molecule poly(ADP-ribose) polymerase (PARP) inhibitor for research use, characterized in the literature for cytoprotective and insulin-sensitizing effects. It is available as a ready-to-use solution or as solid material and is commonly used to probe PARP-related pathways in biochemical and cellular studies.

  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • Demonstrates PARP inhibition with IC50 = 120 μM and Ki = 57 μM.
  • High purity, 98.0% by HPLC.
  • Molecular weight approximately 351.27 g/mol.
  • CAS number 66611-37-8 for unambiguous identification.
  • Suitable for biochemical and cell-based research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.