Learn More
Medchemexpress LLC N-[2-hydroxy-6-(4-methoxybenzamido)phenyl]-4- (4-methyl-1,4-diazepan-1-yl)benzamide | 365462-23-3 | 98.05% | 474.55 | 50 MG
SDP
Catalog No. 5000361590
Encompass_Preferred
Darexaban (YM150) is a potent, selective, and orally active factor Xa (FXa) inhibitor with an IC50 of 54.6 nM. This product exhibits anticoagulant and antithrombotic effects.
  • Shows high selectivity against other related serine proteases, such as trypsin, thrombin, and kallikrein
  • Has anticoagulant and antithrombotic effects
Catalog No. 50-003-61590 Supplier Medchemexpress LLC Supplier No. HY1485350MG
Darexaban (YM150) is a potent, selective, and orally active factor Xa (FXa) inhibitor with an IC50 of 54.6 nM. This product exhibits anticoagulant and antithrombotic effects.
  • Shows high selectivity against other related serine proteases, such as trypsin, thrombin, and kallikrein
  • Has anticoagulant and antithrombotic effects