Learn More
Medchemexpress LLC N-[2-hydroxy-6-(4-methoxybenzamido)phenyl]-4-(4-methyl-1,4-diazepan-1-yl)benzamide | 365462-23-3 | MFCD16620506 | 98.1% | 474.55 | C27H30N4O4 | 10 MG
SDP
Catalog No. 5000286935
Encompass_Preferred
Darexaban is a potent, selective, and orally active factor Xa (FXa) inhibitor used in research to evaluate anticoagulant and antithrombotic effects. It competitively inhibits human FXa (IC50 54.6 nM) and has demonstrated in vivo activity (ED50 24.8 mg/kg in mice), including prolongation of prothrombin time and reduction of thrombus formation in animal models. Supplied as a solid for laboratory research use only.
  • Potent, selective oral factor Xa inhibitor.
  • Competitive inhibition of human FXa with low nanomolar IC50.
  • Demonstrates anticoagulant and antithrombotic activity in animal models.
  • Useful for coagulation and thrombosis study applications.
  • Supplied as a solid with defined storage recommendations for stability.
Catalog No. 50-002-86935 Supplier Medchemexpress LLC Supplier No. HY1485310MG
Darexaban is a potent, selective, and orally active factor Xa (FXa) inhibitor used in research to evaluate anticoagulant and antithrombotic effects. It competitively inhibits human FXa (IC50 54.6 nM) and has demonstrated in vivo activity (ED50 24.8 mg/kg in mice), including prolongation of prothrombin time and reduction of thrombus formation in animal models. Supplied as a solid for laboratory research use only.
  • Potent, selective oral factor Xa inhibitor.
  • Competitive inhibition of human FXa with low nanomolar IC50.
  • Demonstrates anticoagulant and antithrombotic activity in animal models.
  • Useful for coagulation and thrombosis study applications.
  • Supplied as a solid with defined storage recommendations for stability.