Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(2-Hydroxyethyl)ethylenediamine

Catalog No. 1275822KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-758-22KG 2 kg
12-758-2100G 100 g
12-758-21KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-758-22KG Supplier Sigma Aldrich Supplier No. 1275822KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-(2-hydroxyethyl)ethylenediamine (hydeten) or [n-(2-aminoethyl)ethanolamine] can be used as:
  • A precursor in the synthesis of room-temperature ionic liquids, acetate and formate ammonium salts of n-(2-hydroxyethyl)-ethylenediamine
  • A ligand in the preparation of polymeric cyano-bridged platinum(ii)complexes, ligand-copper(ii)carbohydrate complexes and cyano-bridged ni-pt and cu-pt coordination polymers.it can also be used to study the aerobic biodecomposition of amines in hypersaline wastewaters

Specifications

CAS 111-41-1
Density 1.03 g/mL (at 25°C (literature))
Boiling Point 238°C to 240°C (752 mmHg)
Molecular Formula C4H12N2O
Refractive Index n20/D 1.485 (literature)
Linear Formula NH2CH2CH2NHCH2CH2OH
MDL Number MFCD00008170
Quantity 2 kg
Synonym 2-(2-Aminoethylamino)ethanol; N-(2-Aminoethyl)ethanolamine
Molecular Weight (g/mol) 104.15
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.