Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(2-Hydroxyethyl)Ethylenediamine-N,N',N'-Triacetic Acid

Catalog No. H8126500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H8126500G 250 mg
H8126100G 1 kg
H81261KG 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H8126500G Supplier Sigma Aldrich Supplier No. H8126500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(2-Hydroxyethyl)Ethylenediamine-N,N',N'-Triacetic Acid

Specifications

CAS 103404-90-6
Molecular Formula C5H6Na2O5
Linear Formula C5H6O5Na2
Quantity 250 mg
MDL Number MFCD00069573
Synonym N-Carboxymethyl-N'-(2-hydroxyethyl)-N,N'-ethylenediglycine; HEDTA; HEEDTA
Molecular Weight (g/mol) 192.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.