Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)ethylenediaminetriacetic acid, trisodium salt dihydrate, 99+%, Thermo Scientific™

Catalog No. AC171510010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171510010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171510010 Supplier Thermo Scientific Chemicals Supplier No. 171510010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-89-9
Molecular Formula C10H15N2Na3O7
Molecular Weight (g/mol) 344.206
InChI Key WHNXAQZPEBNFBC-UHFFFAOYSA-K
Synonym trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3
PubChem CID 8772
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Quantity 1 kg
Assay Percent Range 99+%
Merck Index 14, 9966
Formula Weight 380.23
Chemical Name or Material N-(2-Hydroxyethyl)ethylenediaminetriacetic acid, trisodium salt dihydrate, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.