Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(2-Hydroxyethyl)Ethylenediaminetriacetic Acid Trisodium Salt Hydrate

Catalog No. 1615353KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-153-53KG 3 kg
16-153-51KG 1 kg
16-153-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-153-53KG Supplier Sigma Aldrich Supplier No. 1615353KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(2-Hydroxyethyl)Ethylenediaminetriacetic Acid Trisodium Salt Hydrate

Specifications

CAS 207386-87-6
Melting Point 288°C
Molecular Formula C10H15N2Na3O7 -+ xH2O
Linear Formula HOCH2CH2N(CH2CO2Na)CH2CH2N(CH2CO2Na)2 -+ xH2O
MDL Number MFCD00150694
Quantity 3 kg
Synonym Trisodium HEDTA hydrate
Molecular Weight (g/mol) 344.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.