Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)hexamethyleneimine, 95%

Catalog No. aal0649706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0649706 5 g
AAL0649714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0649706 Supplier Thermo Scientific Chemicals Supplier No. L0649706
Only null left
Add to Cart

CAS: 20603-00-3 | C8H17NO | 143.23 g/mol

N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

Solubility
Fully miscible in water.

Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20603-00-3
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00020988
InChI Key VMRYMOMQCYSPHS-UHFFFAOYSA-N
Synonym 2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine
PubChem CID 88615
IUPAC Name 2-(azepan-1-yl)ethanol
SMILES C1CCCN(CC1)CCO

Specifications

Density 0.971
Boiling Point 114°C to 115°C (23 mmHg)
Refractive Index 1.484
Quantity 5 g
UN Number UN2735
Beilstein 104110
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 143.22
Percent Purity 95%
Chemical Name or Material N-(2-Hydroxyethyl)hexamethyleneimine
Show More Show Less
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 243-915-2
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.