Learn More
Medchemexpress LLC N-(2-hydroxyethyl)maleimide | 1585-90-6 | MFCD00465266 | 99.2% | 141.12 g/mol | C6H7NO3 | 1 G
SDP
Catalog No. 5000301448
Encompass_Preferred
N-(2-hydroxyethyl)maleimide is a thiol-reactive cross-linker that combines a maleimide reactive group with a hydroxyethyl handle, enabling selective conjugation to thiol-containing molecules and additional chemical modification. It is commonly used in protein labeling, polymer cross-linking, and small-molecule synthesis, and is provided as a high-purity reagent suitable for research applications.
  • Reacts selectively with thiols under mild conditions.
  • Hydroxyethyl group enables further modification and improves solubility.
  • Suitable for protein labeling and bioconjugation workflows.
  • Useful for polymer cross-linking and small-molecule synthesis.
  • High purity supports analytical and synthetic applications.
Catalog No. 50-003-01448 Supplier Medchemexpress LLC Supplier No. HYW0175881G
N-(2-hydroxyethyl)maleimide is a thiol-reactive cross-linker that combines a maleimide reactive group with a hydroxyethyl handle, enabling selective conjugation to thiol-containing molecules and additional chemical modification. It is commonly used in protein labeling, polymer cross-linking, and small-molecule synthesis, and is provided as a high-purity reagent suitable for research applications.
  • Reacts selectively with thiols under mild conditions.
  • Hydroxyethyl group enables further modification and improves solubility.
  • Suitable for protein labeling and bioconjugation workflows.
  • Useful for polymer cross-linking and small-molecule synthesis.
  • High purity supports analytical and synthetic applications.