Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)maleimide 98.0+%, TCI America™
SDP

Catalog No. H15445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15445G 5 g
H15441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15445G Supplier TCI America Supplier No. H15445G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1585-90-6
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00465266
InChI Key AXTADRUCVAUCRS-UHFFFAOYSA-N
Synonym HEMI
PubChem CID 458487
IUPAC Name 1-(2-hydroxyethyl)pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCO

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 141.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.