Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)piperazine 98.5%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
103-76-4
C6H14N2O
130.19
MFCD00005970
WFCSWCVEJLETKA-UHFFFAOYSA-N
n-2-hydroxyethyl piperazine,1-piperazineethanol,1-2-hydroxyethyl piperazine,hydroxyethylpiperazine,2-1-piperazinyl ethanol,2-hydroxyethylpiperazine,2-piperazinoethanol,2-piperazin-1-yl ethan-1-ol,1-piperazinethanol,4-2-hydroxyethyl piperazine
7677
2-piperazin-1-ylethanol
C1CN(CCN1)CCO
103-76-4
C6H14N2O
130.19
MFCD00005970
WFCSWCVEJLETKA-UHFFFAOYSA-N
n-2-hydroxyethyl piperazine,1-piperazineethanol,1-2-hydroxyethyl piperazine,hydroxyethylpiperazine,2-1-piperazinyl ethanol,2-hydroxyethylpiperazine,2-piperazinoethanol,2-piperazin-1-yl ethan-1-ol,1-piperazinethanol,4-2-hydroxyethyl piperazine
7677
2-piperazin-1-ylethanol
C1CN(CCN1)CCO
Density 1.0600g/mL
Refractive Index 1.5055 to 1.5075
Melting Point -38.5°C
Flash Point 135°C
Boiling Point 245°C to 246°C
Specific Gravity 1.06
Color Colorless to Yellow
Quantity 500g
Water 0.5% max.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.50%
Beilstein 23, II, 6
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with most common organic solvents
Formula Weight 130.19
Physical Form Oily Liquid
Percent Purity ≥98.0%
Chemical Name or Material N-(2-Hydroxyethyl)piperazine, 98.5%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-142-3

RTECSNumber : TL6825000

TSCA : TSCA

Recommended Storage : Yellowing of the substance during storage possible