Learn More
Medchemexpress LLC N-2-hydroxyethylpiperazine-N'-ethanesulfonic acid-d18 | 203805-87-2 | MFCD00006158 | 99.9% | 256.42 | C8D18N2O4S | 10 MG
SDP
Catalog No. 5000297475
Encompass_Preferred
Deuterium-labeled HEPES buffer provided as a high-purity solid for analytical and biochemical applications. It serves as a nonvolatile zwitterionic buffering agent effective at pH 6.8-8.2 and is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS work.
  • High isotopic enrichment suitable for NMR and mass spectrometry.
  • High purity (99.9%) for analytical reliability.
  • Effective buffering range pH 6.8-8.2 for biological assays.
  • Available in small milligram quantities for tracer studies.
  • Stable solid form with defined low-temperature storage recommendations.
Catalog No. 50-002-97475 Supplier Medchemexpress LLC Supplier No. HYD0857S10MG
Deuterium-labeled HEPES buffer provided as a high-purity solid for analytical and biochemical applications. It serves as a nonvolatile zwitterionic buffering agent effective at pH 6.8-8.2 and is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS work.
  • High isotopic enrichment suitable for NMR and mass spectrometry.
  • High purity (99.9%) for analytical reliability.
  • Effective buffering range pH 6.8-8.2 for biological assays.
  • Available in small milligram quantities for tracer studies.
  • Stable solid form with defined low-temperature storage recommendations.