Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(2-hydroxyphenyl)picolinamide | 88530-99-8 | 98.0% | 214.22 | C12H10N2O2 | 10 MG
SDP

Catalog No. 5000297897
Encompass_Preferred

N-(2-Hydroxyphenyl)picolinamide is a picolinamide-derived small-molecule research reagent supplied as a solid. It is commonly used as a building block or reference compound in medicinal chemistry and biochemical studies and is provided with accompanying analytical documentation.

  • Purity 98.0%.
  • Molecular weight 214.22 g/mol.
  • Molecular formula C12H10N2O2.
  • Solid, white to off-white appearance.
  • Available in 10 mg, 50 mg, and 100 mg pack sizes.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-97897 Supplier Medchemexpress LLC Supplier No. HY4787510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(2-Hydroxyphenyl)picolinamide is a picolinamide-derived small-molecule research reagent supplied as a solid. It is commonly used as a building block or reference compound in medicinal chemistry and biochemical studies and is provided with accompanying analytical documentation.

  • Purity 98.0%.
  • Molecular weight 214.22 g/mol.
  • Molecular formula C12H10N2O2.
  • Solid, white to off-white appearance.
  • Available in 10 mg, 50 mg, and 100 mg pack sizes.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.