Learn More
Apexbio Technology LLC N-2-HYDROXYPROPYL METHACRYL-1g
SDP
Catalog No. 5000897544
Encompass_Preferred
N-(2-Hydroxypropyl)methacrylamide (CAS No 21442-01-3) is a prominent compound in biomedical research primarily utilized in the field of polymer therapeutics Originating as a derivative of methacrylamide it is well-known for its ability to form biocompatible hydrogels which are frequently used in drug delivery systems The mechanism of action involves the polymerization of the methacrylamide group enabling the encapsulation and controlled release of pharmaceutical agents Its effectiveness is reported in the nanomolar to low micromolar range during in vitro experiments Common applications include studies using cell models animal studies for drug distribution and efficacy evaluation in drug screening processes Experimental concentrations typically depend on the specific research design This compound serves as an essential component for advancing studies in controlled drug release and targeted therapy delivery systems
Catalog No. 50-008-97544 Supplier Apexbio Technology LLC Supplier No. C65931000
N-(2-Hydroxypropyl)methacrylamide (CAS No 21442-01-3) is a prominent compound in biomedical research primarily utilized in the field of polymer therapeutics Originating as a derivative of methacrylamide it is well-known for its ability to form biocompatible hydrogels which are frequently used in drug delivery systems The mechanism of action involves the polymerization of the methacrylamide group enabling the encapsulation and controlled release of pharmaceutical agents Its effectiveness is reported in the nanomolar to low micromolar range during in vitro experiments Common applications include studies using cell models animal studies for drug distribution and efficacy evaluation in drug screening processes Experimental concentrations typically depend on the specific research design This compound serves as an essential component for advancing studies in controlled drug release and targeted therapy delivery systems