Learn More
Medchemexpress LLC N-(2-hydroxypropyl)methacrylamide | 21442-01-3 | MFCD00080531 | 98.9% | 143.18 g/mol | C7H13NO2 | 1 ML
SDP
Catalog No. 5000209362
Encompass_Preferred
N-(2-hydroxypropyl)methacrylamide is a methacrylamide monomer used to synthesize copolymers for polymer-based drug delivery research. Supplied as a ready-to-use 10 mM solution in DMSO for laboratory research use.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Suitable for polymer and copolymer synthesis
  • Applicable to targeted delivery and biomedical research
  • Provided with supporting documentation such as SDS and COA
Catalog No. 50-002-09362 Supplier Medchemexpress LLC Supplier No. HYW07702810MM/1ML
N-(2-hydroxypropyl)methacrylamide is a methacrylamide monomer used to synthesize copolymers for polymer-based drug delivery research. Supplied as a ready-to-use 10 mM solution in DMSO for laboratory research use.
  • Ready-to-use 10 mM solution in DMSO, 1 mL
  • Suitable for polymer and copolymer synthesis
  • Applicable to targeted delivery and biomedical research
  • Provided with supporting documentation such as SDS and COA