Learn More
Medchemexpress LLC N-(2-methoxyphenyl)-N'-[2-(3-pyridinyl)-4-quinazolinyl]-urea | 280570-45-8 | MFCD06691423 | 99.0% | 371.39 | C21H17N5O2 | 5 MG
SDP
Catalog No. 5000247486
Encompass_Preferred
VUF-5574 is a selective antagonist of the adenosine A3 receptor with high affinity (Ki = 4.03 nM). Supplied as a solid white to off-white powder, it is intended for research use in receptor binding and in vitro pharmacology studies.
  • Selective adenosine A3 receptor antagonist (Ki = 4.03 nM).
  • High purity (99.0%) suitable for research applications.
  • Molecular formula C21H17N5O2; molecular weight 371.39.
  • Solid, white to off-white appearance for convenient handling.
  • Available in small research quantities (e.g., 5 mg) for assay development.
Catalog No. 50-002-47486 Supplier Medchemexpress LLC Supplier No. HY1031895MG
VUF-5574 is a selective antagonist of the adenosine A3 receptor with high affinity (Ki = 4.03 nM). Supplied as a solid white to off-white powder, it is intended for research use in receptor binding and in vitro pharmacology studies.
  • Selective adenosine A3 receptor antagonist (Ki = 4.03 nM).
  • High purity (99.0%) suitable for research applications.
  • Molecular formula C21H17N5O2; molecular weight 371.39.
  • Solid, white to off-white appearance for convenient handling.
  • Available in small research quantities (e.g., 5 mg) for assay development.