Learn More
Medchemexpress LLC N-[2-methyl-3-[[4-[4-[[4-(trifluoromethoxy)phenyl]methoxy]-1-piperidinyl]-1,3,5-triazin-2-yl]amino]ph | 1211866-85-1 | MFCD22123242 | 99.7% | 516.52 g·mol⁻¹ | C25H27F3N6O3 | 10 MG
SDP
Catalog No. 5000239803
Encompass_Preferred
TC-N 1752 (CAS 1211866-85-1) is a research-grade small molecule that acts as a potent, orally active inhibitor of Nav1.7 voltage-gated sodium channels. It is supplied as a high-purity solid for use in in vitro and in vivo studies of sodium channel pharmacology and pain signaling.
  • Potent Nav1.7 inhibitor with reported submicromolar activity.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.7%) appropriate for research applications.
  • Stable when stored as powder at -20°C or 4°C under recommended conditions.
  • Molecular weight 516.52 g·mol⁻¹ and formula C25H27F3N6O3.
Catalog No. 50-002-39803 Supplier Medchemexpress LLC Supplier No. HY10740510MG
TC-N 1752 (CAS 1211866-85-1) is a research-grade small molecule that acts as a potent, orally active inhibitor of Nav1.7 voltage-gated sodium channels. It is supplied as a high-purity solid for use in in vitro and in vivo studies of sodium channel pharmacology and pain signaling.
  • Potent Nav1.7 inhibitor with reported submicromolar activity.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.7%) appropriate for research applications.
  • Stable when stored as powder at -20°C or 4°C under recommended conditions.
  • Molecular weight 516.52 g·mol⁻¹ and formula C25H27F3N6O3.