Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[2-methyl-3-[[4-[4-[[4-(trifluoromethoxy)phenyl]methoxy]piperidin-1-yl]-1,3,5-triazin-2-yl]amino]phen | 1211866-85-1 | MFCD22123242 | 99.7% | 516.52 g/mol | C25H27F3N6O3 | 1 ML
SDP

Catalog No. 5000241539
Encompass_Preferred

TC-N 1752 is a small-molecule sodium channel inhibitor used in research to probe Nav1.7 activity. It is provided as a ready-to-use solution suitable for in vitro pharmacology and electrophysiology studies.

  • Potent Nav1.7 sodium channel inhibitor.
  • Supplied as a 10 mM solution in DMSO in a 1 mL vial.
  • High purity (99.7%) suitable for assay work.
  • Molecular weight 516.52 g/mol; chemical formula C25H27F3N6O3.
  • CAS number 1211866-85-1 for unambiguous identification.

Catalog No. 50-002-41539 Supplier Medchemexpress LLC Supplier No. HY10740510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC-N 1752 is a small-molecule sodium channel inhibitor used in research to probe Nav1.7 activity. It is provided as a ready-to-use solution suitable for in vitro pharmacology and electrophysiology studies.

  • Potent Nav1.7 sodium channel inhibitor.
  • Supplied as a 10 mM solution in DMSO in a 1 mL vial.
  • High purity (99.7%) suitable for assay work.
  • Molecular weight 516.52 g/mol; chemical formula C25H27F3N6O3.
  • CAS number 1211866-85-1 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.