Learn More
Medchemexpress LLC N-[2-methyl-3-[[4-[4-[[4-(trifluoromethoxy)phenyl]methoxy]piperidin-1-yl]-1,3,5-triazin-2-yl]amino]phenyl]acetamide | 1211866-85-1 | MFCD22123242 | 99.7% | 516.52 g/mol | C25H27F3N6O3 | 25 MG
SDP
Catalog No. 5000239943
Encompass_Preferred
TC-N 1752 is a small-molecule research compound that inhibits Nav1.7 (voltage-gated sodium) channels and is used in pharmacology and electrophysiology studies of sodium channel function and pain signaling. The substance is identified by CAS 1211866-85-1, has molecular formula C25H27F3N6O3 and a molecular weight of about 516.5 g/mol. It is supplied as a solid with DMSO solubility and is accompanied by analytical data supporting its stated purity.
  • Potent Nav1.7 inhibitory activity useful for sodium channel research.
  • High reported purity suitable for biochemical assays.
  • Characterized chemical identity with CAS and molecular formula available.
  • Soluble in DMSO for preparation of experimental solutions.
  • Available in multiple pack sizes to accommodate different study scales.
Catalog No. 50-002-39943 Supplier Medchemexpress LLC Supplier No. HY10740525MG
TC-N 1752 is a small-molecule research compound that inhibits Nav1.7 (voltage-gated sodium) channels and is used in pharmacology and electrophysiology studies of sodium channel function and pain signaling. The substance is identified by CAS 1211866-85-1, has molecular formula C25H27F3N6O3 and a molecular weight of about 516.5 g/mol. It is supplied as a solid with DMSO solubility and is accompanied by analytical data supporting its stated purity.
  • Potent Nav1.7 inhibitory activity useful for sodium channel research.
  • High reported purity suitable for biochemical assays.
  • Characterized chemical identity with CAS and molecular formula available.
  • Soluble in DMSO for preparation of experimental solutions.
  • Available in multiple pack sizes to accommodate different study scales.