Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Methylbutyryl)glycine, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187895 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4444 10 mg
11-101-4445 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4444 Supplier MilliporeSigma Supelco Supplier No. 0447810MG

Specifications

CAS 52320-67-9
Linear Formula C7H13 NO3
MDL Number MFCD09736911
Synonym 2-Methylbutyryl glycine; N-(2-Methyl-1-oxobutyl)glycine
Type Analytical Standard
Quantity 10 mg
Formula Weight 159.18
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.