Learn More
Medchemexpress LLC N-[2-[N'-pentyl-(6,6-dimethyl-2,4-heptadiynyl)amino]ethyl]- (2-methyl-1-naphthylthio)acetamide | 138046-43-2 | 100.0% | 452.63 g·mol⁻¹ | C31H36N2O | 50 MG
SDP
Catalog No. 5000239848
Encompass_Preferred
YM-750 is a small-molecule inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) with a reported IC50 of 0.18 μM. Supplied as a high-purity solid, it is used in biochemical and cell-based research to investigate cholesterol esterification and cellular lipid metabolism.
  • Potent ACAT inhibitor (IC50 = 0.18 μM) for biochemical assays.
  • High purity suitable for research (reported 99.95%).
  • Solid powder formulation for accurate weighing and handling.
  • Storage recommendations provided for long-term stability (powder: -20°C).
  • Available in multiple research-scale pack sizes, including 50 mg.
  • Applicable to studies of cholesterol esterification and lipid metabolism.
Catalog No. 50-002-39848 Supplier Medchemexpress LLC Supplier No. HY10739650MG
YM-750 is a small-molecule inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) with a reported IC50 of 0.18 μM. Supplied as a high-purity solid, it is used in biochemical and cell-based research to investigate cholesterol esterification and cellular lipid metabolism.
  • Potent ACAT inhibitor (IC50 = 0.18 μM) for biochemical assays.
  • High purity suitable for research (reported 99.95%).
  • Solid powder formulation for accurate weighing and handling.
  • Storage recommendations provided for long-term stability (powder: -20°C).
  • Available in multiple research-scale pack sizes, including 50 mg.
  • Applicable to studies of cholesterol esterification and lipid metabolism.