Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-2-Nitrophenylsulfenyl-L-valine Dicyclohexylammonium Salt, TCI America™
SDP

Catalog No. N01645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N01645G Supplier TCI America Supplier No. N01645G

Chemical Identifiers

CAS 7675-57-2
Molecular Formula C23H37N3O4S
Molecular Weight (g/mol) 451.626
MDL Number MFCD00038494
InChI Key PIXHCLWIWCGYKF-RLGBJRSSSA-N
Synonym N-(2-Nitrophenylthio)-L-valine Dicyclohexylammonium Salt, N-Nps-L-valine Dicyclohexylammonium Salt, Nps-Val-OH.DCHA
PubChem CID 91974102
IUPAC Name N-cyclohexylcyclohexanamine;(2R)-3-methyl-2-[(2-nitrophenyl)sulfanylamino]butanoic acid
SMILES CC(C)C(C(=O)O)NSC1=CC=CC=C1[N+](=O)[O-].C1CCC(CC1)NC2CCCCC2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 451.63
Physical Form Crystalline Powder
Chemical Name or Material N-2-Nitrophenylsulfenyl-L-valine Dicyclohexylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.