Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Phenylethyl)acetamide 98.0+%, TCI America™
SDP

Catalog No. P106625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P106625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P106625G Supplier TCI America Supplier No. P106625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 877-95-2
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00026177
InChI Key MODKMHXGCGKTLE-UHFFFAOYSA-N
Synonym n-phenethylacetamide, n-2-phenylethyl acetamide, n-acetyl-2-phenylethylamine, n-acetylphenethylamine, acetamide, n-2-phenylethyl, n-phenethyl acetamide, n-acetylphenylethylamine, n-beta-phenylethylacetamide, acetamide, n-phenethyl, unii-2jxy218szi
PubChem CID 70143
ChEBI CHEBI:18177
IUPAC Name N-(2-phenylethyl)acetamide
SMILES CC(=O)NCCC1=CC=CC=C1

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Phenylethyl)acetamide
RTECSNumber AC7740000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.