Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[2-(tert-Butoxycarbonylamino)ethyl]glycine Ethyl Ester 95.0+%, TCI America™
SDP

Catalog No. B36321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36321G Supplier TCI America Supplier No. B36321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72648-80-7
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.307
InChI Key WVFXXOLKYIPCAX-UHFFFAOYSA-N
Synonym N-[2-(Boc-amino)ethyl]glycine Ethyl Ester
PubChem CID 4319362
IUPAC Name ethyl 2-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethylamino]acetate
SMILES CCOC(=O)CNCCNC(=O)OC(C)(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 246.31
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[2-(tert-Butoxycarbonylamino)ethyl]glycine Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.