Learn More
Cayman Chemical N 2AzIdoethylbetuLNamIde 10mg
SDP
Catalog No. 502946326
Encompass_Preferred
A pentacyclic triterpenoid intermediate; an intermediate in the synthesis of betulonic acid derivatives with in vitro cancer cell cytotoxicity
Catalog No. 50-294-6326 Supplier Cayman Chemical Supplier No. 3306110 MG
A pentacyclic triterpenoid intermediate; an intermediate in the synthesis of betulonic acid derivatives with in vitro cancer cell cytotoxicity