Learn More
Cayman Chemical N 2phenylethylIndomethacI 10mg
SDP
Catalog No. 502895619
Encompass_Preferred
N-2PIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-2PIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.06 and 0.125 µM, respectively.{8243,8513} It is over 400 times less potent as an inhibitor of ovine COX-1.{8513}
Catalog No. 50-289-5619 Supplier Cayman Chemical Supplier No. 7027210 MG
N-2PIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-2PIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.06 and 0.125 µM, respectively.{8243,8513} It is over 400 times less potent as an inhibitor of ovine COX-1.{8513}