Learn More
Cayman Chemical N-(2-phenylethyl)-Indomethacin amide, Purity: ≥98%, CAS: 261766-32-9, 50 mg
SDP
Catalog No. 502895563
Encompass_Preferred
N-2PIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-2PIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.06 and 0.125 µM, respectively.{8243,8513} It is over 400 times less potent as an inhibitor of ovine COX-1.{8513}
Catalog No. 50-289-5563 Supplier Cayman Chemical Supplier No. 7027250 MG
N-2PIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-2PIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.06 and 0.125 µM, respectively.{8243,8513} It is over 400 times less potent as an inhibitor of ovine COX-1.{8513}