Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(2R)-4-amino-3,4-dioxo-1-phenylbutan-2-yl]-4-(2-fluorophenyl)-2-methyl-1,3-oxazole-5-carboxamide | 2221010-42-8 | 98.0% | 395.38 | C21H18FN3O4 | 5 MG
SDP

Catalog No. 5000262950
Encompass_Preferred

Dazcapistat is a research-grade calpain inhibitor with reported IC50s of <3 μM for calpain 1, calpain 2, and calpain 9. Supplied as a white to off-white solid with a stated purity of 98.0%, it is intended for in vitro and in vivo research; follow supplier protocols for solubility, formulation, and storage.

  • Molecular formula C21H18FN3O4; molecular weight 395.38
  • Purity 98.0%
  • Appearance white to off-white solid
  • In vitro solubility: DMSO 100 mg/mL (ultrasonic recommended)
  • In vivo formulation: ≥2.5 mg/mL in recommended vehicle protocols (10% DMSO/40% PEG300/5% Tween-80/45% saline; alternatives available)
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-62950 Supplier Medchemexpress LLC Supplier No. HY1328505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dazcapistat is a research-grade calpain inhibitor with reported IC50s of <3 μM for calpain 1, calpain 2, and calpain 9. Supplied as a white to off-white solid with a stated purity of 98.0%, it is intended for in vitro and in vivo research; follow supplier protocols for solubility, formulation, and storage.

  • Molecular formula C21H18FN3O4; molecular weight 395.38
  • Purity 98.0%
  • Appearance white to off-white solid
  • In vitro solubility: DMSO 100 mg/mL (ultrasonic recommended)
  • In vivo formulation: ≥2.5 mg/mL in recommended vehicle protocols (10% DMSO/40% PEG300/5% Tween-80/45% saline; alternatives available)
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.