Learn More
Medchemexpress LLC N-[(2R)-4-amino-3,4-dioxo-1-phenylbutan-2-yl]-4-(2-fluorophenyl)-2-methyl-1,3-oxazole-5-carboxamide | 2221010-42-8 | 98.0% | C21H18FN3O4 | 10MG
SDP
Catalog No. 5000261493
Encompass_Preferred
Dazcapistat is a small-molecule calpain inhibitor with reported IC50s of <3 μM for calpain 1, calpain 2, and calpain 9. It is provided as a high-purity research reagent in both powder and solution formats for biochemical and cell-based studies, with recommended storage conditions to maintain stability.
  • Potent calpain inhibition with IC50s <3 μM for calpain 1, 2, and 9.
  • High purity (98.0%) suitable for research applications.
  • Available as powder and pre-made solution formats for flexible use.
  • Defined storage recommendations to preserve compound integrity.
  • Appropriate for biochemical assays and cellular studies of protease activity.
Catalog No. 50-002-61493 Supplier Medchemexpress LLC Supplier No. HY13285010MG
Dazcapistat is a small-molecule calpain inhibitor with reported IC50s of <3 μM for calpain 1, calpain 2, and calpain 9. It is provided as a high-purity research reagent in both powder and solution formats for biochemical and cell-based studies, with recommended storage conditions to maintain stability.
  • Potent calpain inhibition with IC50s <3 μM for calpain 1, 2, and 9.
  • High purity (98.0%) suitable for research applications.
  • Available as powder and pre-made solution formats for flexible use.
  • Defined storage recommendations to preserve compound integrity.
  • Appropriate for biochemical assays and cellular studies of protease activity.