Learn More
Medchemexpress LLC N-[3-(1,3-benzothiazol-2-yl)-6-propan-2-yl-5,7-dihydro-4H-thieno[2,3-c]pyridin-2-yl]acetamide | 524708-03-0 | MFCD11984329 | 99.5% | C19H21N3OS2 | 10MG
SDP
Catalog No. 5000261794
Encompass_Preferred
APE1-IN-1 (CAS 524708-03-0) is a small-molecule inhibitor of apurinic/apyrimidinic endonuclease 1 (APE1) that is reported to penetrate the blood-brain barrier and to inhibit APE1 activity in vitro (IC50 ≈ 2 μM). It is provided as a research reagent and has been used in cellular studies to increase the cytotoxicity of alkylating agents.
Catalog No. 50-002-61794 Supplier Medchemexpress LLC Supplier No. HY13673110MG
APE1-IN-1 (CAS 524708-03-0) is a small-molecule inhibitor of apurinic/apyrimidinic endonuclease 1 (APE1) that is reported to penetrate the blood-brain barrier and to inhibit APE1 activity in vitro (IC50 ≈ 2 μM). It is provided as a research reagent and has been used in cellular studies to increase the cytotoxicity of alkylating agents.