Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[3-(1H-indol-4-yl)-5-(2-methoxypyridine-4-carbonyl)phenyl]methanesulfonamide | 915412-67-8 | MFCD18782727 | 99.9% | C22H19N3O4S | 10MG
SDP

Catalog No. 5000261632
Encompass_Preferred

LP-261 is an orally active antimitotic small-molecule research compound that inhibits tubulin polymerization and has demonstrated in vivo efficacy against human non-small-cell lung tumor xenografts. It is supplied as a high-purity solid and is documented with analytical files for research use.

  • Inhibits tubulin polymerization (EC50 3.2 μM).
  • Demonstrated in vivo antitumor activity in xenograft models.
  • High purity solid with available COA, LC-MS, and SDS documentation.
  • Molecular formula C22H19N3O4S; molecular weight 421.47 g·mol⁻¹.
  • Soluble in DMSO (≈33.33 mg/mL) for stock solution preparation.
  • Storage recommendations provided for powder and solution formats.

Catalog No. 50-002-61632 Supplier Medchemexpress LLC Supplier No. HY1438910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LP-261 is an orally active antimitotic small-molecule research compound that inhibits tubulin polymerization and has demonstrated in vivo efficacy against human non-small-cell lung tumor xenografts. It is supplied as a high-purity solid and is documented with analytical files for research use.

  • Inhibits tubulin polymerization (EC50 3.2 μM).
  • Demonstrated in vivo antitumor activity in xenograft models.
  • High purity solid with available COA, LC-MS, and SDS documentation.
  • Molecular formula C22H19N3O4S; molecular weight 421.47 g·mol⁻¹.
  • Soluble in DMSO (≈33.33 mg/mL) for stock solution preparation.
  • Storage recommendations provided for powder and solution formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.