Learn More
Medchemexpress LLC N-(3-(1H-pyrazol-5-yl)phenyl)-2-oxo-6-(trifluoromethyl)-1,2-dihydropyridine-3-carboxamide | 00-00-0 | 98.0% | C16H11F3N4O2 | 10MG
SDP
Catalog No. 5000260495
Encompass_Preferred
NRX-1532 is a small-molecule research compound that enhances the interaction between β-catenin and β-TrCP and is used to probe Wnt/β-catenin signaling in cellular and biochemical studies.
  • Enhances β-catenin:β-TrCP interaction for signaling studies.
  • High purity (98.0%) suitable for research applications.
  • Soluble in DMSO at 250 mg/mL for in vitro use.
  • Solid form, stable when stored as powder at -20°C.
  • In solvent, stable up to 6 months at -80°C and 1 month at -20°C.
  • Offered in multiple small-scale solid and solution pack formats for convenience.
Catalog No. 50-002-60495 Supplier Medchemexpress LLC Supplier No. HY14498210MG
NRX-1532 is a small-molecule research compound that enhances the interaction between β-catenin and β-TrCP and is used to probe Wnt/β-catenin signaling in cellular and biochemical studies.
  • Enhances β-catenin:β-TrCP interaction for signaling studies.
  • High purity (98.0%) suitable for research applications.
  • Soluble in DMSO at 250 mg/mL for in vitro use.
  • Solid form, stable when stored as powder at -20°C.
  • In solvent, stable up to 6 months at -80°C and 1 month at -20°C.
  • Offered in multiple small-scale solid and solution pack formats for convenience.