Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(3-((1R)-1-((6R)-4-Hydroxy-2-oxo-6-((1R)-2-(phenyl-d5)ethyl-1,2-d2)-6-propyl-5,6-dihydro-2H-pyran- | 174484-41-4 | 98.9% | 609.71 g/mol | C31H26D7F3N2O5S | 10 MG
SDP

Catalog No. 5000286477
Encompass_Preferred

Tipranavir-d7 is a deuterium-labeled isotopologue of an HIV-1 protease inhibitor provided as a high-purity research standard for analytical and tracer studies. It is intended for use as an internal standard in LC-MS, pharmacokinetic experiments, and method development where isotopic labeling improves quantitation and compound tracking.

  • Deuterated (d7) isotopic label suitable for tracer and internal standard use.
  • High isotopic enrichment for improved mass discrimination.
  • High purity: 98.9% (HPLC).
  • Molecular weight 609.71 g/mol and formula C31H26D7F3N2O5S.
  • Supplied as a small mass (10 MG) solid for analytical workflows.
  • Appropriate for LC-MS method development and quantitative studies.

Catalog No. 50-002-86477 Supplier Medchemexpress LLC Supplier No. HY15148S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tipranavir-d7 is a deuterium-labeled isotopologue of an HIV-1 protease inhibitor provided as a high-purity research standard for analytical and tracer studies. It is intended for use as an internal standard in LC-MS, pharmacokinetic experiments, and method development where isotopic labeling improves quantitation and compound tracking.

  • Deuterated (d7) isotopic label suitable for tracer and internal standard use.
  • High isotopic enrichment for improved mass discrimination.
  • High purity: 98.9% (HPLC).
  • Molecular weight 609.71 g/mol and formula C31H26D7F3N2O5S.
  • Supplied as a small mass (10 MG) solid for analytical workflows.
  • Appropriate for LC-MS method development and quantitative studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.