Learn More
Medchemexpress LLC N-[3-[(3,4-dihydro-2,4-dioxo-3-propylthieno[3,2-d]pyrimidin-1(2H)-yl)methyl]phenyl]-benzeneacetamide | 2098776-03-3 | 98.4% | C24H23N3O3S | 10MG
SDP
Catalog No. 5000260013
Encompass_Preferred
FEN1-IN-SC13 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) used for preclinical research into DNA replication, repair, and antitumor effects. It inhibits FEN1 activity in vitro and in cells, impairing DNA processing and increasing sensitivity to DNA-damaging agents. Supplied as a powder with high reported purity for laboratory research use only.
  • Potent FEN1 inhibition in vitro and in cells.
  • Interferes with DNA replication and repair pathways.
  • Demonstrated antitumor activity in xenograft models.
  • High purity suitable for biochemical and cellular assays.
  • Powder form with recommended low-temperature storage.
Catalog No. 50-002-60013 Supplier Medchemexpress LLC Supplier No. HY14575810MG
FEN1-IN-SC13 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) used for preclinical research into DNA replication, repair, and antitumor effects. It inhibits FEN1 activity in vitro and in cells, impairing DNA processing and increasing sensitivity to DNA-damaging agents. Supplied as a powder with high reported purity for laboratory research use only.
  • Potent FEN1 inhibition in vitro and in cells.
  • Interferes with DNA replication and repair pathways.
  • Demonstrated antitumor activity in xenograft models.
  • High purity suitable for biochemical and cellular assays.
  • Powder form with recommended low-temperature storage.