Learn More
Medchemexpress LLC N-[[3-[[3-[(5-chlorothiophen-2-yl)sulfonylamino]-4-methoxyindazol-1-yl]methyl]phenyl]methyl]-2-hydrox | 1240516-71-5 | MFCD30533311 | 99.8% | 549.06 g/mol | C24H25ClN4O5S2 | 100 MG
SDP
Catalog No. 5000215478
Encompass_Preferred
GSK2239633A is a small-molecule CCR4 antagonist used in research to inhibit [125I]-TARC binding to human CCR4 (reported pIC50 7.96). It is supplied as a high-purity solid with defined storage recommendations and high solubility in DMSO for assay preparation.
  • High purity suitable for research assays.
  • Selective CCR4 antagonistic activity (pIC50 7.96).
  • Solid form with recommended freezer storage.
  • High solubility in DMSO for stock preparation.
  • Molecular weight 549.06 g/mol and formula C24H25ClN4O5S2.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-15478 Supplier Medchemexpress LLC Supplier No. HY100183100MG
GSK2239633A is a small-molecule CCR4 antagonist used in research to inhibit [125I]-TARC binding to human CCR4 (reported pIC50 7.96). It is supplied as a high-purity solid with defined storage recommendations and high solubility in DMSO for assay preparation.
  • High purity suitable for research assays.
  • Selective CCR4 antagonistic activity (pIC50 7.96).
  • Solid form with recommended freezer storage.
  • High solubility in DMSO for stock preparation.
  • Molecular weight 549.06 g/mol and formula C24H25ClN4O5S2.
  • For research use only; not for human or diagnostic use.