Learn More
Medchemexpress LLC N-(3-((3-(5-chlorothiophene-2-sulfonamido)-4-methoxyindazol-1-yl)methyl)benzyl)-2-hydroxy-2-methylpropanamide | 1240516-71-5 | 99.8% | 549.06 | C24H25ClN4O5S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10018325MG
Encompass_Preferred
GSK2239633A is a small-molecule CC-chemokine receptor 4 (CCR4) antagonist supplied for research use. It inhibits [125I]-TARC binding to human CCR4 with a reported pIC50 of 7.96 and shows activity in human CD4+ CCR4+ T-cells. The compound is provided as a solid with high reported purity and defined molecular formula and weight, suitable for in vitro pharmacology and analytical studies.
  • Allosteric CCR4 antagonist with reported pIC50 ~7.96.
  • Validated in vitro activity on human CD4+ CCR4+ T-cells.
  • High reported purity suitable for research assays.
  • Solid form, stable for analytical use.
  • Contains defined molecular formula and molecular weight for characterization.
Catalog No. 50-002-15736