Learn More
Medchemexpress LLC N-[[3-(4-acetylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide | 104421-21-8 | 98.0% | 276.29 g/mol | C14H16N2O4 | 5 MG
SDP
Catalog No. 5000221387
Encompass_Preferred
DuP-721 is a broad-spectrum, orally active oxazolidinone antibacterial agent with in vitro activity against susceptible and drug-resistant Mycobacterium tuberculosis (typical inhibitory concentrations 1.5-4 μg/mL) and demonstrated protective efficacy in mouse infection models.
  • Broad-spectrum antibacterial activity, including efficacy against drug-resistant M. tuberculosis.
  • In vitro inhibitory concentrations typically 1.5-4 μg/mL against M. tuberculosis.
  • Demonstrated oral protective efficacy in mouse infection models.
  • Molecular formula C14H16N2O4 and molecular weight 276.29 g/mol.
  • High purity (about 98.0%) as supplied.
  • Solid, white to off-white physical form with recommended cold storage.
Catalog No. 50-002-21387 Supplier Medchemexpress LLC Supplier No. HY1396185MG
DuP-721 is a broad-spectrum, orally active oxazolidinone antibacterial agent with in vitro activity against susceptible and drug-resistant Mycobacterium tuberculosis (typical inhibitory concentrations 1.5-4 μg/mL) and demonstrated protective efficacy in mouse infection models.
  • Broad-spectrum antibacterial activity, including efficacy against drug-resistant M. tuberculosis.
  • In vitro inhibitory concentrations typically 1.5-4 μg/mL against M. tuberculosis.
  • Demonstrated oral protective efficacy in mouse infection models.
  • Molecular formula C14H16N2O4 and molecular weight 276.29 g/mol.
  • High purity (about 98.0%) as supplied.
  • Solid, white to off-white physical form with recommended cold storage.