Learn More
Medchemexpress LLC N-(3-(4-amino-7-(2-deoxy-pentofuranosyl)-prop-2-yn-1-yl)-2,2,2-trifluoroacetamide | 178420-75-2 | MFCD29904589 | 98.8% | 399.32 g·mol⁻¹ | C16H16F3N5O4 | 5 MG
SDP
Catalog No. 5000221677
Encompass_Preferred
7-TFA-ap-7-Deaza-dA is a modified deoxynucleoside that contains an alkyne functional group for bioorthogonal labeling. It is intended for incorporation into DNA or RNA and for conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC), supplied as a well-characterized solid or as a DMSO solution for synthesis and labeling workflows.
  • High purity of 98.8% for reliable synthetic use.
  • Alkyne handle enables CuAAC click chemistry conjugation.
  • Compatible with DNA and RNA synthesis protocols.
  • Available as solid or as 10 mM solution in DMSO for convenience.
  • Characterized molecular weight and formula for analytical confirmation.
Catalog No. 50-002-21677 Supplier Medchemexpress LLC Supplier No. HY1385905MG
7-TFA-ap-7-Deaza-dA is a modified deoxynucleoside that contains an alkyne functional group for bioorthogonal labeling. It is intended for incorporation into DNA or RNA and for conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC), supplied as a well-characterized solid or as a DMSO solution for synthesis and labeling workflows.
  • High purity of 98.8% for reliable synthetic use.
  • Alkyne handle enables CuAAC click chemistry conjugation.
  • Compatible with DNA and RNA synthesis protocols.
  • Available as solid or as 10 mM solution in DMSO for convenience.
  • Characterized molecular weight and formula for analytical confirmation.