Learn More
Medchemexpress LLC N-[3,5-bis(trifluoromethyl)phenyl]-1,2-benzisothiazol-3-amine 1,1-dioxide | 823830-85-9 | 99.7% | 394.30 Da | C15H8F6N2O2S | 5 MG
SDP
Catalog No. 5000221374
Encompass_Preferred
M1002 is a small-molecule allosteric agonist of hypoxia-inducible factor-2 (HIF-2) that enhances expression of HIF-2 target genes. It is supplied as a solid research reagent for in vitro and biochemical studies to modulate HIF-2 signaling pathways.
  • High chemical purity (99.7%).
  • Well-defined molecular formula C15H8F6N2O2S and molecular weight 394.30 Da.
  • Identified by CAS 823830-85-9 for unambiguous reference.
  • Recommended storage: powder -20°C (stable up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for cellular and biochemical assays to activate HIF-2 signaling.
Catalog No. 50-002-21374 Supplier Medchemexpress LLC Supplier No. HY1392875MG
M1002 is a small-molecule allosteric agonist of hypoxia-inducible factor-2 (HIF-2) that enhances expression of HIF-2 target genes. It is supplied as a solid research reagent for in vitro and biochemical studies to modulate HIF-2 signaling pathways.
  • High chemical purity (99.7%).
  • Well-defined molecular formula C15H8F6N2O2S and molecular weight 394.30 Da.
  • Identified by CAS 823830-85-9 for unambiguous reference.
  • Recommended storage: powder -20°C (stable up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for cellular and biochemical assays to activate HIF-2 signaling.