Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide | 978-62-1 | MFCD00218820 | 99.3% | 383.67 g/mol | C15H8ClF6NO2 | 1 ML
SDP

Catalog No. 5000245385
Encompass_Preferred

IMD-0354 (IKK2 Inhibitor V) is a small-molecule inhibitor of IKKβ (IKK2) that blocks NF-κB signaling and downstream NF-κB-dependent transcription. It is intended for research use and is available as a solid or as a prepared 10 mM solution in DMSO.

  • Selective IKKβ inhibitor that reduces NF-κB activity.
  • Reported to inhibit TNF-α-induced NF-κB transcription with an IC50 of ~1.2 μM.
  • High purity of approximately 99.3% suitable for research applications.
  • Molecular weight 383.67 g/mol for accurate dosing calculations.
  • Soluble in DMSO; supplier recommends DMSO at high concentration.
  • Available as solid material or as a 10 mM solution in DMSO for convenience.

Catalog No. 50-002-45385 Supplier Medchemexpress LLC Supplier No. HY1017210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IMD-0354 (IKK2 Inhibitor V) is a small-molecule inhibitor of IKKβ (IKK2) that blocks NF-κB signaling and downstream NF-κB-dependent transcription. It is intended for research use and is available as a solid or as a prepared 10 mM solution in DMSO.

  • Selective IKKβ inhibitor that reduces NF-κB activity.
  • Reported to inhibit TNF-α-induced NF-κB transcription with an IC50 of ~1.2 μM.
  • High purity of approximately 99.3% suitable for research applications.
  • Molecular weight 383.67 g/mol for accurate dosing calculations.
  • Soluble in DMSO; supplier recommends DMSO at high concentration.
  • Available as solid material or as a 10 mM solution in DMSO for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.