Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-[3,5-Bis(Trifluoromethyl)Phenyl]-N'-[(8A,9S)-6'-Methoxy-9-Cinchonanyl]Thiourea

Catalog No. 690481250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-048-1250MG 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 69-048-1250MG Supplier Sigma Aldrich Supplier No. 690481250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-[3,5-Bis(trifluoromethyl)phenyl]-N'sa-[(8a,9S)-6'sa-methoxy-9-cinchonanyl]thiourea is a bifunctional cinchona organocatalyst, which can be used to synthesize: Stereoselective diaryl(nitro)butanone via enantioselective Michael addition of nitromethane to chalcones. Enantioselective +a-amino acids via asymmetric Mannich reaction of malonates with aryl and alkyl imines. The synthesis of 3-indolylmethanamines by the reaction of indoles with imines via asymmetric Friedel-Crafts reaction. The enantioselective conjugate addition of active methylene compounds to enones to obtain the corresponding addition products.

Specifications

CAS 951021-12-8
Molecular Formula C5H5F6N3O6S2
Linear Formula C5H5F6N3O6S2
Refractive Index n20/D 1.4275
MDL Number MFCD11656053
Quantity 50 g
Molecular Weight (g/mol) 381.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.