Learn More
Strem, An Ascensus Company CAS# 1326239-56-8. 50mg. N-[3,5-Bis(trifluoromethyl)phenyl]-N'-[(9R)-6'-methoxycinchonan-9-yl]urea, 95%, (99% ee)
SDP
Supplier:  Strem, An Ascensus Company 07632750MG
Encompass
CAS# 1326239-56-8. 50mg. N-[3,5-Bis(trifluoromethyl)phenyl]-N'-[(9R)-6'-methoxycinchonan-9-yl]urea, 95%, (99% ee). Molecular Weight: 578.50. Molecular Formula: C29H28F6N4O2. Color/form: white-light yellow pwdr. Strem# 07-6327. http://www.strem.com/catalog/v/07-6327/
Catalog No. NC2085030