Learn More
Medchemexpress LLC N'-[3-[[5-bromo-2-[3-(pyrrolidine-1-carbonylamino)anilino]pyrimidin-4-yl]amino]propyl]-2,2-dimethylpropanediamide | 702676-93-5 | MFCD12922516 | 99.4% | 547.45 g·mol⁻1 | C23H31BrN8O3 | 1 MG
SDP
Catalog No. 5000243934
Encompass_Preferred
BX-320 is a selective, ATP-competitive, orally active small-molecule inhibitor of PDK1 for preclinical research. It inhibits PDK1 with an IC50 of 30 nM, blocks PDK1/Akt signaling, induces apoptosis in tumor cells, and has demonstrated antiproliferative and antitumor activity in cell-based and animal models.
  • Selective ATP-competitive PDK1 inhibitor (IC50 30 nM).
  • Blocks PDK1/Akt signaling and induces apoptosis in tumor cells.
  • Demonstrates antiproliferative activity in cell assays and efficacy in in vivo tumor models.
  • Exhibits off-target activity against Chk1, c-Kit, KDR, PKA, CDK2, GSK3β, and PKC at micromolar concentrations.
  • High purity (99.4%).
  • Molecular weight 547.45 g·mol⁻1; formula C23H31BrN8O3.
  • Store solid at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-43934 Supplier Medchemexpress LLC Supplier No. HY105151MG
BX-320 is a selective, ATP-competitive, orally active small-molecule inhibitor of PDK1 for preclinical research. It inhibits PDK1 with an IC50 of 30 nM, blocks PDK1/Akt signaling, induces apoptosis in tumor cells, and has demonstrated antiproliferative and antitumor activity in cell-based and animal models.
  • Selective ATP-competitive PDK1 inhibitor (IC50 30 nM).
  • Blocks PDK1/Akt signaling and induces apoptosis in tumor cells.
  • Demonstrates antiproliferative activity in cell assays and efficacy in in vivo tumor models.
  • Exhibits off-target activity against Chk1, c-Kit, KDR, PKA, CDK2, GSK3β, and PKC at micromolar concentrations.
  • High purity (99.4%).
  • Molecular weight 547.45 g·mol⁻1; formula C23H31BrN8O3.
  • Store solid at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).