Learn More
Medchemexpress LLC N-[3-[[5-chloro-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]thio]phenyl]-2-propenamide | 1214265-57-2 | 99.2% | 5 MG
SDP
Catalog No. 5000234411
Encompass_Preferred
WZ8040 is an irreversible, mutant-selective inhibitor of the epidermal growth factor receptor (EGFR) with activity against the T790M resistance mutation. It inhibits EGFR phosphorylation and is used as a laboratory research compound to study mutant EGFR signaling and inhibitor sensitivity.
  • Irreversible inhibitor of EGFR T790M.
  • Demonstrates substantially greater potency against mutant EGFR compared with wild-type.
  • Available as solid powder and as DMSO solutions for ready-to-use applications.
  • Typical purity around 99.2% as supplied.
  • Soluble in DMSO (high solubility reported) and formulatable for in vivo dosing protocols.
  • Store powder at -20°C for long-term stability; store stock solutions at -80°C when possible.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-34411 Supplier Medchemexpress LLC Supplier No. HY120295MG
WZ8040 is an irreversible, mutant-selective inhibitor of the epidermal growth factor receptor (EGFR) with activity against the T790M resistance mutation. It inhibits EGFR phosphorylation and is used as a laboratory research compound to study mutant EGFR signaling and inhibitor sensitivity.
  • Irreversible inhibitor of EGFR T790M.
  • Demonstrates substantially greater potency against mutant EGFR compared with wild-type.
  • Available as solid powder and as DMSO solutions for ready-to-use applications.
  • Typical purity around 99.2% as supplied.
  • Soluble in DMSO (high solubility reported) and formulatable for in vivo dosing protocols.
  • Store powder at -20°C for long-term stability; store stock solutions at -80°C when possible.
  • Intended for research use only; not for human or veterinary use.