Learn More
Medchemexpress LLC N-(3-(5-chloro-2-(difluoromethoxy)phenyl)-1-(2-(4-((2-cyanoethyl)(methyl)amino)piperidin-1-yl)-2-oxoet | 1831144-46-7 | 99.9% | 612.03 | C28H28ClF2N9O3 | 10MG
SDP
Catalog No. 5000238744
Encompass_Preferred
iJak-381 is a selective JAK1/2 inhibitor for laboratory research, used to study Janus kinase-mediated cytokine signaling and inflammatory responses. It has reported activity against IL-13, IL-4, and IL-6 pathways and has been evaluated in preclinical studies for lung-restricted inhalation delivery.
  • Selective JAK1/2 inhibitor for research applications.
  • Blocks IL-13, IL-4, and IL-6 signaling pathways.
  • Lung-restricted profile suitable for respiratory disease models.
  • High purity for reproducible experimental results.
  • Supplied as a 10 mg research sample.
  • Molecular formula C28H28ClF2N9O3, molecular weight 612.03.
Catalog No. 50-002-38744 Supplier Medchemexpress LLC Supplier No. HY12501910MG
iJak-381 is a selective JAK1/2 inhibitor for laboratory research, used to study Janus kinase-mediated cytokine signaling and inflammatory responses. It has reported activity against IL-13, IL-4, and IL-6 pathways and has been evaluated in preclinical studies for lung-restricted inhalation delivery.
  • Selective JAK1/2 inhibitor for research applications.
  • Blocks IL-13, IL-4, and IL-6 signaling pathways.
  • Lung-restricted profile suitable for respiratory disease models.
  • High purity for reproducible experimental results.
  • Supplied as a 10 mg research sample.
  • Molecular formula C28H28ClF2N9O3, molecular weight 612.03.